Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eisco LabGlass® Distilling Flasks with 3 Angled Necks, Round Bottom, Interchangable Joints
Supplier: Eisco Scientific
CH0418T CH0418O CH0418P CH0418M CH0418V CH0418K CH0418L CH0418N CH0418R CH0418U CH0418Q CH0418S
470332-140EA 48.3 USD InStock
470332-140 470332-142 470332-130 470332-144 470332-122 470332-132 470332-124 470332-134 470332-126 470332-136 470332-128 470332-138
Eisco LabGlass® Distilling Flasks with 3 Angled Necks, Round Bottom, Interchangable Joints
Flasks Distillation Flasks
High quality round bottom distilling flasks with 3 angles necks.
  • Made of high quality borosilicate glass 3.3
  • Interchangable, ground joints
  • 3 angled neck design
  • Round bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty