Would you like to log in again?
Second Law of Motion Apparatus
Supplier: EISCO SCIENTIFIC LLC
PH0319PRBL PH0319
470332-228EA 12.4 USD InStock
470332-228 470230-116
Second Law of Motion Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Metal apparatus designed to demonstrate second law of motion.
  • This Second Law of Motion apparatus is made of solid metal and designed to last many years in the class room
  • Base is painted blue and is 10 x 2.5"
  • Spring driven and offers two tensions
  • Includes 7.5" x 0.50" dia. rod to be able to mount on table clamp or fixed to a support stand
  • Complete with two steel balls
This Newton's Second Law of Motion Apparatus is made of solid metal and designed to last many years in the class room. This is spring driven heavy to demonstrates Newton's Second law of Motion. The unit is based on a substantial metal casting and the entire mechanism is visible. The spring gun offers two tensions. The 7.5" long unit has a 0.5" diameter rod so it may be horizontally clamped to a support stand or table clamp. Complete with two steel balls ( One drilled ) This shows that both balls will hit the floor at the same time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty