Would you like to log in again?
Eisco® Vibration Generator
Supplier: EISCO SCIENTIFIC LLC
VBGN
470332-242EA 194.95 USD InStock
470332-242
Eisco® Vibration Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Useful for physics and sound demonstrations.
  • 7 to 8 mm maximum peak to peak displacement at 1 Hz, decreasing with increase in frequency
  • 1 to 5 KHz total frequency range
  • 3 Ω at 50 Hz coil impedance:
  • Vibration generator can be used in any position, free standing or clamped
The vibration generator is basically a loud speaker. A coil wound on a thin tube moves within a permanent magnetic field when current of an altering nature is applied. The frequency response encompasses the whole of the audio spectrum and beyond.
Electrical input is made via two 4 mm sockets and the mechanical output is provided by a shaft. The unit gives mechanical oscillations when fed by signals from an oscillator / A.F. amplifier or Power Signal Generator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty