Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sterile Microcentrifuge and Centrifuge Tubes
Supplier: MTC BIO, INC.
C3150-S C2625-R C2600 C2230 C2625-B C2605 C2602 C2410 C2630
470332-390BG 20.4 USD InStock
470332-390 470332-380 470332-394 470332-384 470332-392 470332-382 470332-388 470332-386 470332-378
Sterile Microcentrifuge and Centrifuge Tubes
Tubes Centrifuge Tubes
These sterile tubes come in a variety of sizes and packaging options to fulfill a variety of tube needs in the lab.
  • RNAse-, DNAse-, pyrogen-, and cytotoxin-free
  • Store at −80 °C
  • Sterilized by gamma radiation
These centrifuge tubes are perfect for everyday sample preparation needs. The rugged construction provides durable, reliable, long-term sample storage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty