Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Miniature DC Motor, with Alligator Leads, United Scientific

Supplier: United Scientific Supplies
DCM045-L
470332-540EA 4.95 USD InStock
470332-540
Miniature DC Motor, with Alligator Leads, United Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Miniature DC motor designed for experiments using low starting voltage (4.5 V) and current (70 mA).

  • Speed: 1000 - 4900 rpm
  • Motor approximately 34mm L x 24mm Dia.
  • Shaft is 10mm L x 2mm Dia.

Includes soldered-on 7" lead with alligator clips.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty