Would you like to log in again?
Fischertechnik TXT 4.0 IOT Add-On Pack
Supplier: STUDICA INC.
Fischertechnik
559897
470332-596EA 182.29 USD InStock
470332-596
Fischertechnik TXT 4.0 IOT Add-On Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Professional-level entry into environmental sensing and the Internet of things.
  • Pair with the TXT 4.0 Base set
  • Teaching Topics Include: measurement acquisition and transmission, network connections, cloud computing, IoT
  • Suitable for Middle/High School students
  • Downloadable teaching and activity guides
Professional entry into measured value acquisition with the Robotics ADD-ON IoT! Together with the Robotics TXT 4.0 Base set, the sensor station enables the measurement of temperature, humidity, air pressure, air quality and brightness. The sensor station can be programmed and controlled with the ROBO Pro Coding programming software and the ROBOTICS TXT 4.0 Controller and is ideal for teaching topics such as measurement acquisition and transmission as well as control and regulation of actuators and sensors. The measured value acquisition is carried out via the connection of the TXT 4.0 Controller with the fischertechnik cloud, in which the sensor data is stored, collected and graphically displayed. Via the user interface, the so-called dashboard, the various sensor data are permanently recorded (real-time) and the camera, which can be swiveled in two axes, is remotely controlled. The teaching material created to accompany the Robotics ADD-ON IoT set contains 6 experiments and associated solutions.
Ordering information: Required but available separately: TXT 4.0 Base set.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty