Would you like to log in again?
Economy Magnet Kit
Supplier: United Scientific Supplies
MGTKIT
470332-606EA 13.15 USD InStock
470332-606
Economy Magnet Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
An exceptional value.
  • Includes a number of different magnets
  • Great for teaching
This kit includes a variety of magnets and accessories for teaching basics of magnetism. Includes a reusable storage box and an activity guide.
Kit includes the following: 1 steel horseshoe magnet, 3 ceramic donut magnets, 1 iron filings pack, 1 lodestone, 1 activity guide containing 17 experiments, 2 ceramic bar magnets, 4 metal squares (iron, copper, aluminum, and zinc), and 2 magnetic compasses.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty