Would you like to log in again?
Bimetal Strip/Compound Bar, United Scientific
Supplier: United Scientific Supplies
BMST05
470332-608EA 4.05 USD InStock
470332-608
Bimetal Strip/Compound Bar, United Scientific
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Simple tool to explore the thermal properties of matter.
  • Bimetal strip is composed of two alloys that display different responses when exposed to heat
  • Strips are securely bonded and fastened to a sturdy wooden handle
  • Activity guide included
A quick and easy tool for demonstrating thermal properties of matter. Bar is bi-metallic and has a wooden handle attached. When heated, the bar bends dramatically.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty