Would you like to log in again?
Super Spring with Stand
Supplier: United Scientific Supplies
SSPRING
470332-610EA 21.45 USD InStock
470332-610
Super Spring with Stand
Educational Materials Physics Educational Materials Mechanics Learning Activities
Huge wave demonstrations.
  • Extra-long spring
  • Includes activity guide
Extra-long spring (21cm when compressed) allows users to magnify the wave demonstrations to illustrate traveling waves and wave patterns. Included stand prevents tangling or damage to super spring during storage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty