Would you like to log in again?
Liquid Convection Apparatus, United Scientific
Supplier: United Scientific Supplies
UNLCA001
470332-614EA 42.2 USD InStock
470332-614
Liquid Convection Apparatus, United Scientific
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
The liquid convection apparatus is designed to show the convection of heat in a liquid.
  • 20 mm Ext.Ø. glass tube
  • Tube is bent into 200×150 mm rectangle
The liquid convection apparatus is made from borosilicate glass, and is bent into a rectangle with a filling funnel. To show the convection of heat in a liquid, simply fill it with a colored liquid and place a heat source at one of the bottom corners. Glass tube is bent into a rectangle. Comes in a custom package to ensure product can be stored safely.
Required but not included are a support stand, clamp, heat source, and colored liquid.
Packaging: Supplied in custom package.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty