Would you like to log in again?
Dynamics Carts, United Scientific
Supplier: United Scientific Supplies
DNCART2
470332-618EA 19.25 USD InStock
470332-618
Dynamics Carts, United Scientific
Educational Materials Physics Educational Materials Mechanics Learning Activities
Set of two dynamics carts to demonstrate energy conservation, velocity, elastic collisions, acceleration, and momentum.
  • Designed with durable spring steel bumpers and rubber stoppers
  • Features convenient wells to accommodate additional masses
  • Activity guide included
Ideal for demonstrating experiments in conservation, velocity, elastic collisions, acceleration, and momentum. Included are two impact carts and two spring steel bumpers with rubber stopper assemblies. Each cart features a well for additional masses (not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty