Would you like to log in again?
Advanced Thermostat Kit
Supplier: United Scientific Supplies
UNADTHK01 UNADTHK02
470332-624EA 289.6 USD InStock
470332-624 470332-622
Advanced Thermostat Kit
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Learn how home thermostats and circuit breakers operate.
  • Students can see and investigate the relationship between parallel circuits and currents
  • Individual and Class sets available
Observe energy transfer and conversion and investigate the systems that keep homes comfortable and safe with this versatile apparatus. Guided activities help students explore the relationship between parallel circuits and current, understand how thermostats and circuit breakers operate, and compare the energy efficiency of LEDs to incandescent bulbs. Includes activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty