Would you like to log in again?
Van De Graaff Generator, Small Electric, United Scientific
Supplier: United Scientific Supplies
VANDGF1-SEB VANDGF1-SE
470332-628EA 349.45 USD InStock
470332-628 470332-630
Van De Graaff Generator, Small Electric, United Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This excellent quality electrostatic generator consists of a charge generation system, an aluminum collector sphere, and an electric motor drive on a sturdy base.
  • Requires a 110 VAC outlet
  • Includes activity guide
The charge generation system is based on a moving neoprene belt mounted on insulating rollers with metal charge transfer combs. The system is housed in a clear tube that allows for easy viewing of the moving parts. A 100,000 volt potential can be generated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty