Would you like to log in again?
Advanced Projection Ripple Tank, United Scientific
Supplier: United Scientific Supplies
RPTANK1
470332-632EA 773.3 USD InStock
470332-632
Advanced Projection Ripple Tank, United Scientific
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
The Advanced Projection Ripple Tank is a comprehensive system for demonstrating basic wave phenomena using water waves.
  • Variable frequency generator
  • Large vertical display
  • Bright LED lights
A horizontal glass plate is mounted in a water-tight frame with foam beaches around the perimeter to suppress unwanted wave reflections. This tank is mounted on three tall legs with an angled mirror beneath the glass plate. A translucent screen is fitted to the front of the apparatus to display the projected image. A stray light shield can be placed around the back and sides of the apparatus to prevent ambient light from reaching the screen. In use, a pool of water a few millimeters deep is filled into the tank. A ripple generator for mounting and energizing ripplers is fitted to one side of the tank and an LED stroboscope lamp is adjustably mounted above the glass plate on a gooseneck support to provide the projection light. The ripple generator and the LED stroboscope lamp are both driven by a Wave Monitor control unit that synchronizes the wave generator’s oscillations with the stroboscope flashes to produce a stationary projected image. The wave generator can also be operated asynchronously, allowing the stroboscope flash frequency to be varied while the wave generator frequency remains fixed. This generates a moving projected image of variable speed. The included accessory set contains a series of five different ripplers (two of each), a pair of acrylic blocks, a barrier with slits, a clear foil with a printed grid, a plastic dropper, and a metric ruler.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty