Would you like to log in again?
Center Groove, Double-Bevel 12" Ruler, United Scientific
Supplier: United Scientific Supplies
SCLW12
470332-646EA 1.25 USD InStock
470332-646
Center Groove, Double-Bevel 12" Ruler, United Scientific
Rulers
This 12" (30.5 cm) wooden ruler has a double bevel and a pencil groove.
  • Features double bevel and pencil groove
  • Metal edge on one side
Ruler is scaled in 1/16" increments on one bevel, and 1 mm increments on the opposite bevel. There is a metal edge on one side.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty