Would you like to log in again?
UniStore™ Large Reagent Bottles, Narrow Mouth, PP
Supplier: United Scientific Supplies
UniStore™
BNMP2L-PK/6 BNMP2L BNMP8L BNMP4L BNMP4L-PK/6 BNMP8L-PK/6
470332-680EA 20.75 USD InStock
470332-680 470332-690 470332-684 470332-682 470332-688 470332-686
UniStore™ Large Reagent Bottles, Narrow Mouth, PP
Bottles Plastic Bottles
Large size reagent bottles.
  • Autoclavable
UniStore™ narrow mouth PP large capacity reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L). Constructed of polypropylene, the bottles are translucent, pliable, and have excellent strength for extended use. Ideal for packing, shipping, and storage purposes.
Ordering information: Lined HDPE cap for a tight seal included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty