Would you like to log in again?
Mini Weather Station, United Scientific
Supplier: United Scientific Supplies
MWS136
470332-724EA 9.55 USD InStock
470332-724
Mini Weather Station, United Scientific
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
Introduce basic weather concepts with this versatile apparatus.
  • Device freely rotates to indicate wind direction
  • Removable rain gauge allows for the quick assessment of rainfall
Measure temperature (in Fahrenheit and Celsius) and wind speed, then reference the screen-printed chart to easily calculate wind chill. The product is designed to mount on posts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty