Would you like to log in again?
Earth Sun Planetarium, United Scientific
Supplier: United Scientific Supplies
UNSLSY02
470332-726EA 71.95 USD InStock
470332-726
Earth Sun Planetarium, United Scientific
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
Demonstrate the rotational motion of Earth on its axis, its orbit around the Sun, and the revolution of the moon around Earth with this high-quality model.
  • Battery operated sun
  • Moon rotates around the Earth
  • Illustrates the different seasons
As the Sun casts a beam of light onto Earth (via an embedded LED), users can easily manipulate the positions of each body to examine how their dynamics correspond to seasons, eclipses, and lunar phases.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty