Would you like to log in again?
Newton's Ring Apparatus, United Scientific
Supplier: United Scientific Supplies
NEWRIN2
470332-746EA 19.95 USD InStock
470332-746
Newton's Ring Apparatus, United Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
The Newton’s Rings Apparatus consists of a plano-convex lens on top of a flat glass plate, held together by a plastic frame.
  • Easy to adjust thumbscrews
  • Demonstrate optical properties of lenses
The pressure between the lens and the plate can be adjusted by means of three thumbscrews.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty