Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LED Stroboscope, United Scientific

Supplier: United Scientific Supplies
STROBE01
470332-750EA 350.4 USD InStock
470332-750
LED Stroboscope, United Scientific
Educational Materials Physics Educational Materials Optics Learning Activities
Hand-held stroboscope with 30W super-LED delivers bright flashes at adjustable rates from 1 to 250 Hz.

  • Display modes: Frequency (Hz or rpm)
  • Frequency range: 1.0 to 250.0 Hz; 1 to 15000 rpm
  • Frequency resolution: 0.1 Hz; 1 rpm
  • Frequency adjustment modes: frequency doubling (X2), frequency halving (÷2), incremental by 0.1 Hz/1 RPM (+, –)
  • Bulb power: 30 W LED
  • Dimensions: 18×6×4 cm
  • Battery life: 3 hours continuous use

This unique hand-held stroboscope was made possible by the development of high-power LEDs. Ideal for the motion rates encountered in lab situations. Features magnetic pads on back for attaching to vertical steel surfaces. Requires four AAA batteries (not included).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty