Would you like to log in again?
United Scientific Hall's Carriage
Supplier: United Scientific Supplies
United Scientific
HLCRG1 HLCRG1-USA USA001
470332-768EA 9.55 USD InStock
470332-768 470340-064 470332-766
United Scientific Hall's Carriage
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Sturdy, smooth rolling dynamics cart, with universal smartphone adapter.
  • Classroom Hall's carriage designed for use with all types of inclined planes
  • Durable plastic body with machined aluminum wheels
  • Roomy well for additional masses (masses not included)
  • Utilize the power of a smartphone with the optional adapter
This Hall's carriage features machined aluminum wheels and axles mounted on hardened steel cone bearings for minimum friction and long life. For use with all types of inclined planes.
The unique adapter holds a smartphone securely for motion experiments. Using an app of their own choosing, users can graph acceleration and velocity.
Delivery information: Includes activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty