Would you like to log in again?
Loop The Loop Track, United Scientific
Supplier: United Scientific Supplies
LOOP01
470332-770EA 39.2 USD InStock
470332-770
Loop The Loop Track, United Scientific
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Ideal for classroom demonstrations of centripetal acceleration and transformation of potential energy into kinetic energy.
  • Includes activity guide
The Loop the Loop Track demonstrates centripetal acceleration and the transformation of potential energy into kinetic energy. Included is a sturdy base with rubber feet, metal ramp with loop, 16 mm steel ball, and 19 mm steel ball. The metal ramp with loop features a net at the end of the loop to receive the steel ball.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty