Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

United Scientific Spark Timer

Supplier: United Scientific Supplies
United Scientific
SPARK1
470332-772EA 67.95 USD InStock
470332-772
United Scientific Spark Timer
Timers Mechanical Timers
The spark timer is used for measuring velocity and acceleration of falling or moving objects.

  • Spark timer device
  • 60 Hz (mains power frequency)
  • Tape length: 45 m
  • Tape width: 17 mm

A spark timer is used for measuring velocity and acceleration of falling or moving objects. When the tape is pulled through the timer a spark is created which burns tiny dots in the tape at a precise and same interval. The faster the tape is pulled through the timer the larger the distance between the dots. This allows students to calculate the acceleration. Spark tape, carbon disks, weight and clamp are included. Includes activity guide.

Delivery information: Timer includes five carbon disks, one 45 m roll of tape, clamp, and weight.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty