Would you like to log in again?
United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
Supplier: United Scientific Supplies
PDB030
470332-794EA 1.45 USD InStock
470332-794
United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
Bottles Dropping and Dispensing Bottles
This dropper bottle is made of translucent and low density polyethylene.
  • For use when dispensing small drops of a solution
  • Drop sizes are uniform for consistent dispensing of liquids
Bottle can be used with or without dropper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty