Would you like to log in again?
Room Divider, 6' Height, 7 Sections, Clear Acrylic Panels,
National Public Seating
Supplier: NATIONAL PUBLIC SEATING SE
RDB6-7CA
470332-924EA 3581.95 USD InStock
470332-924
Room Divider, 6' Height, 7 Sections, Clear Acrylic Panels,
National Public Seating
Prefabricated Rooms
Divide your multipurpose room with portable, lightweight, extremely durable, acrylic Robo™ dividers.
  • NPS clear acrylic dividers are built with ³/₁₆" clear OPTIX® acrylic
  • These clear panels are lightweight, extremely durable, resistant to intense force or shock and are non-yellowing
  • NPS acrylic panels are completely encased and sealed in aluminum frames
  • Self-leveling casters
  • Full-length hinges
  • Unique foot-pedal release
  • Heavyweight- non tipping
Divide your multipurpose room, medical center or house of worship with the ultimate partition: the durable, flexible and portable Robo™ dividers. Room to room transportation is made simple with full length handles and spring enhanced casters that glide right over thresholds and uneven flooring. They feature Tech-Lock brakes at both ends for a secure stop. Our portable dividers are made with steel, aluminum-framed panels and can be ganged together with magnet connectors built into the divider ends. Dividers are available in PET and clear acrylic.
They stand 6' tall and come in four lengths. Go green with the environmentally-friendly PET divider, made with 100% recycled materials. The PET panels dampen sound, so outside distractions won't stand a chance. Or go with our clear acrylic Robo™ dividers to see and hear it all while maintaining a distance. The acrylic panels are crafted with lightweight, non-yellowing OPTIX® acrylic, a highly durable material that resists intense force or shock. Robo™ - Robust. Easy. Portable.
Certifications: ANSI-BIFMA Certified and MAS Certified Green.
Ordering information: Two-week Quickship. Ten-year warranty. PET dividers are available separately.
Delivery information: Shipped fully assembled
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty