Would you like to log in again?
Tamper Guard® Evidence Tape
Supplier: FORENSICS SOURCE
BD2100
470334-306EA 33 USD InStock
470334-306
Tamper Guard® Evidence Tape
Educational Materials Forensics Educational Materials Forensics Supplies
Makes any container tamperproof.
  • Easily secures to containers
  • Shows evidence of physical or chemical tampering
  • Write-on
Virtually impossible to violate, TAMPER GUARD® Evidence Tape outperforms all evidence tapes and is ideal for anchoring and sealing today’s exotic plastics and other finishes. Each roll is printed in safety orange using a special dye that will bleed and smear if it comes into contact with most solvents, and is printed on a thin acrylic stock that tears easily. A superior adhesive and minute saw-tooth edges along both sides of the tape leave signs of tampering. Easy to write on, this tape is ideal for sealing evidence or for secure storage of nearly any item.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty