Would you like to log in again?
Evidence Pillbox
Supplier: FORENSICS SOURCE
3-3000
470334-386PK 137.9 USD InStock
470334-386
Evidence Pillbox
Educational Materials Forensics Educational Materials Forensics Supplies
Protective storage for evidence.
  • Easily store fragile evidence
  • Base slides out of top
Ideal for carrying and storing small items of evidence. Pack evidence with a small amount of cotton to prevent evidence from moving and becoming damaged during handling or transit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty