Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Disposable Transfer Pipettes
Supplier: G.S.C. INTERNATIONAL, INC.
25008-100 25025-100 25013 25025 25013-100 25008 25009-100 25007 25007-100 25023-100
470334-422PK 17.95 USD InStock
470334-422 470334-412 470334-420 470334-410 470334-408 470334-406 470334-418 470334-416 470334-424 470334-414
Disposable Transfer Pipettes
Pipettes Pasteur/Transfer Pipettes
Used in many applications such as urinalysis, wet chemistry, microbiology, hematology, and serology.
  • Small micro-tip
  • Made from LDPE
Ensure a clean, contamination-free experience every time. Whether working in a lab, a medical facility, or an industrial setting, this is an essential tool for STEM professionals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty