Would you like to log in again?
LDPE Pasteur Pipettes, United Scientific Supplies
Supplier: United Scientific Supplies
P31206 P31205 P31202 P31201
470334-444PK 31.8 USD InStock
470334-444 470334-442 470334-440 470334-446
LDPE Pasteur Pipettes, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Non-toxic All-in-one pipettes.
  • Molded
  • Unbreakable
  • Translucent
  • Pack of 500
These pipettes are all non-toxic and molded from unbreakable translucent lowdensity polyethylene. These can be sealed and refrigerated. They are available in sterile and non-sterile forms. The sterile pipettes are individually wrapped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty