Would you like to log in again?
Special Use Forceps
Supplier: AFTAB AHMED SE
14-1010 15-TW088 15-122 14-0520 14-0512 14-0590
470334-840BX 243.9 USD InStock
470334-840 470334-772 470334-784 470334-782 470334-814 470334-812
Special Use Forceps
Forceps
Specialty forceps for student use.
  • Stainless steel
  • A variety of options available
One set of forceps isn't like every other. This selection provides options for many users. Tweezers are a basic sharp point forceps with a very short blade length that makes the ends very easy to control and provides a stronger grip.
Cover slipping forceps are designed with a broader smooth tip that is bent at an angle to improve applying a coverslip to a microscope slide.
Microdissection forceps are similar to a watchmakers forceps with a lighter weight blade, requiring less force to close. Easy grip forceps have an enlarged grip loop part-way down each blade to provide additional stability and ease of closure for a narrow blade.
Finally the self closing forceps act in a reverse motion to most. Squeezing opens the jaws, and they close when released. These are easier to use if long-term holding is required.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty