Would you like to log in again?
Watchmaker Forceps
Supplier: AFTAB AHMED SE
14-0505 14-1400
470334-804BX 48.2 USD InStock
470334-804 470334-802
Watchmaker Forceps
Forceps
Ultra-fine point forceps.
  • Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty