Would you like to log in again?
Surgical Style Scissors
Supplier: AFTAB AHMED SE
10-785 14-0981 14-0990 14-09882 10-788 10-790 14-0988 10-798 10-787
470334-884BX 63.05 USD InStock
470334-884 470334-874 470334-882 470334-850 470334-880 470334-858 470334-834 470334-886 470334-876
Surgical Style Scissors
Scissors
Multiple sizes and styles.
  • Stainless steel
Multiple styles of surgical scissors for dissection. Use Sharp pointed scissors to start a dissection cut without a scalpel, and the blunt sides inside the specimen to minimize the chances of accidental internal damage.
The stainless construction will cut tougher tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty