Would you like to log in again?
Dissection Knives
Supplier: AFTAB AHMED SE
14-1190 14-0930 14-0969
470334-852BX 103.5 USD InStock
470334-852 470334-890 470334-844
Dissection Knives
Knives
Great for rough dissection.
  • Stainless steel construction
  • Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty