Would you like to log in again?
Fixed Blade Scalpels
Supplier: AFTAB AHMED SE
14-4320 14-0875 14-0860
470334-896BX 355.85 USD InStock
470334-896 470334-808 470334-826
Fixed Blade Scalpels
Scalpels and Scalpel Assemblies Scalpels
More durable than replaceable blades.
  • Pre-sharpened
Fixed blade scalpels are more durable than replaceable blades. With less flex there is less likelihood of blade breakage during a dissection.
Best used for more coarse dissection near hard tissue, these offer three different materials of sharpening.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty