Would you like to log in again?
Wards® Red Cabbage Juice pH Indicator
Supplier: ALDON CORP SE
Wards®
IS5068
470334-898EA 17.5 USD InStock
470334-898
Wards® Red Cabbage Juice pH Indicator
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
A colorful way to test the pH of various solutions!
  • Comes with 20 g of red cabbage powder and instructions
Learn about acids, bases, and pH indicators with our red cabbage juice kit. A pH indicator is a chemical that behaves differently depending on the pH level it is exposed to. Red cabbage juice is a natural pH indicator that changes color from red to purple to yellow-green as the pH of a solution increases. Test out different substances to determine if they are acidic or basic, and see if you can make all six colors found on the color chart!
Delivery information: 20g Red cabbage powder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty