Would you like to log in again?
United Scientific Supplies Specimen Jars, Screw-Cap, Glass
Supplier: GSC INTERNATIONAL SE
410-3 410-4-70-400 410-5 410-2
470334-908EA 4.65 USD InStock
470334-908 470334-906 470334-912 470334-910
United Scientific Supplies Specimen Jars, Screw-Cap, Glass
Jars Specimen Jars
Flint glass specimen jars for easy storage.
  • Autoclavable
All the wide mouth jars are made from soda lime glass and have a PP cap with a PE liner. Since the opening is so large, it is easy to insert and remove materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty