Would you like to log in again?
Wards® Introduction to Basic, Selective, and Differential Media
Supplier: ALDON CORP SE
Wards®
IS3610
470336-152EA 197.9 USD InStock
470336-152
Wards® Introduction to Basic, Selective, and Differential Media
Educational Materials Biology Educational Materials Microbiology Learning Activities
Learn about the different types of media used to grow bacteria.
  • Kit contains enough materials for 10 groups
In this activity, students will learn about the different types of media that can be used to grow bacteria. They will predict which media will support the growth of different bacteria, then streak the different media with bacteria cultures to observe their growth. The bacteria growth will be evaluated to determine if students' predictions were correct
Delivery information: Includes 2×200 ml prepared Tryptic soy agar; 2×200 ml prepared MacConkey agar; 10 lyophilized cultures of Serratia marcescens; 10 lyophilized cultures of Bacillus subtilis;
10 lyophilized cultures of Escherichia coli; 30 Tryptic soy broth tubes; 40 sterile Petri dishes; 30 sterile pipettors; 40 Cotton swab packs, 2/pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty