Would you like to log in again?
Wards® Introduction to Streak Isolation and Aseptic Technique
Supplier: ALDON CORP SE
Wards®
IS3611
470336-154EA 98.95 USD InStock
470336-154
Wards® Introduction to Streak Isolation and Aseptic Technique
Educational Materials Biology Educational Materials Microbiology Learning Activities
Indroduce skills that microbiologists use on a daily basis.
  • Learn about different types of media and how bacteria reproduce
In this activity, students will be introduced to the skills and techniques that microbiologists use on a daily basis. Students will practice these techniques by performing a streak isolation with the provided bacterial cultures, as well as samples they collect from their classroom, to learn about different types of media and how bacterium reproduce.
Delivery information: Include 2×200 ml prepared Tryptic soy agar; 10 lyophilized cultures of Serratia marcescens; 20 sterile Petri dishes; 10 Tryptic soy broth tubes; 10 Inoculation loops; 10 Sterile pipettors;
10 Sterile cotton swabs, pack/2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty