Would you like to log in again?
Wards® DNA Extraction Lab Activity
Supplier: ALDON CORP SE
Wards®
IS3002-SGL
470336-174EA 23.2 USD InStock
470336-174
Wards® DNA Extraction Lab Activity
Educational Materials Biology Educational Materials Genetics Learning Activities
Use biological detergents, enzymes, and ethanol to isolate DNA from plant material.
  • Teacher's manual and study guide are included
In this lab you will learn the history of the discovery of DNA and DNA structure. Understand the nature of genetic inheritance and the role of DNA and proteins in genetic expression while using biological detergents, enzymes, and ethanol to isolate DNA from plant material. You need to supply the plant material. Kit contains enough materials for 15 groups. Teacher’s Manual and Student Study Guide copymasters are included.
Delivery information: Includes 250 ml 7.5% SDS/1.5% NaCl; 20 ml Ethanol, 95%; 1 Pepsin (to make 25 ml 0.5% solution); 10 Zipper bags; 5 Filters; 5 Plastic tubes; 10 Graduated pipettes; 5 Stirrers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty