Would you like to log in again?
Wards® Equilibrium and LeChatlier's Principle
Supplier: ALDON CORP SE
Wards®
IS2526
470336-178EA 30.35 USD InStock
470336-178
Wards® Equilibrium and LeChatlier's Principle
Educational Materials Chemistry Educational Materials Thermochemistry/Kinetics Learning Activities
Investigate equilibrium and LeChatlier's principle through 4 different experiments.
  • Kit contains enough materials for 15 groups
  • Teacher's guide and student study guide copymasters are included
Students will investigate equilibrium and Le Chatelier’s principle through four different experiments. They will apply various stresses to different chemical systems at equilibrium, then use their understanding of Le Chatelier’s principle to explain their results. Kit contains enough material for 15 groups. Teacher’s Guide and Student Study Guide copymasters are included.
Delivery information: Includes 25 ml Bromothymol blue; 50 ml Sodium hydroxide, 0.1 M; 25 ml Hydrochloric acid, 0.1 M; 150 ml Potassium thiocyanate solution; 25 ml Ferric nitrate solution; 15 g Potassium thiocyanate crystals; 15 g Sodium phosphate crystals; 150 ml Copper (II) sulfate solution; 100 ml Ammonium hydroxide, 1M; 25 ml Hydrochloric acid, 1M; 50 ml Methyl red.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty