Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HOBO® Data Loggers Self-Describing 6' Stainless Steel Temperature Sensor
Supplier: ONSET COMPUTER CORPORATION SE
SD-TEMP-SS-06
470336-314EA 132.29 USD InStock
470336-314
HOBO® Data Loggers Self-Describing 6' Stainless Steel Temperature Sensor
Data Loggers
Stainless steel temperature sensor with Self-Describing technology for quick, easy set up.
  • Locking jack to ensure sensor is connected and stay connected
  • Food-grade stainless-steel temperature probe
  • Available in 1.8 m (6')
For use with HOBO MX1104 and MX1105 multi-channel data loggers, this food-grade stainless-steel temperature sensor offers integrated strain relief, so the probe is secured to the logger. It features Self-Describing technology to automatically communicate configuration information to the logger, without any programming or extensive user setup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty