Would you like to log in again?
12-Well Plastic Spot Plates, Eisco Scientific
Supplier: Eisco Scientific
CH0542CPK12
470336-548PK 14.55 USD InStock
470336-548
12-Well Plastic Spot Plates, Eisco Scientific
Staining Tiles
Durable and reusable spot plates for microchemistry and general laboratory use.
  • Made from high quality, durable polyethylene
  • Each plate contains 12 individual wells
  • Wells measure approximately 1" in diameter
  • Wells have a capacity of approximately 1.5 ml
  • Easy to wash and reuse
  • Great classroom tool for containing and observing small-scale reactions
High quality polyethylene well spotting plates used for observing small chemical reactions. The plastic material these plates are made from allows them to be washed and reused. Can be used to compare results among students in a lab, or in classroom demonstrations.
Packaging: Spot plates are supplied in packs of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty