Would you like to log in again?
Pipetting By Numbers: STEAM Pipetting Practice, Edvotek
Supplier: Edvotek
S-45
470336-716KT 64.45 USD InStock
470336-716
Pipetting By Numbers: STEAM Pipetting Practice, Edvotek
Educational Materials Laboratory Safety Educational Materials
Help your students master the fundamental biotechnology technique of micro pipetting while creating a colorful, science-themed classroom poster as well as their own artwork.
  • Set your classroom up for a year of experimental success by mastering micro pipetting
  • Learn best practices, key parts, and the importance of accuracy and precision
  • Engage students with a STEAM wet lab based on paint by numbers
  • Create a colorful, science-themed classroom poster
This kit includes materials for 10 lab groups and takes approximately 45 minutes to complete.
Ordering information: Materials required but not included: automatic micropipettes with tips, small containers for discarding used tips, scissors, and tape.
Materials included: instructions, dyes, test strips, Pipetting by Numbers canvas and blank canvas, microcentrifuge tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty