Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cryogenic Vials, MTC Bio
Supplier: MTC Bio
V4805 V3801 V3802 V3805 V4802 V3803 V4803 V4801
470336-828CS 207.5 USD InStock
470336-828 470336-826 470336-838 470336-832 470336-830 470336-824 470336-836 470336-834
Cryogenic Vials, MTC Bio
Vials
These cryogenic vials come sterile in convenient resealable bags of 50 vials.
  • Self-standing star foot design
  • Gamma sterilized
  • Rnase, Dnase, pyrogen, and endotoxin free
  • Free locking rack included with each case
Made of durable LN₂ grade polypropylene, these vials can withstand temperatures from –196 to +121 °C. The O-ring on the inner cap is made with silicone to prevent leakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty