Would you like to log in again?
Medicor® Peripheral Intravenous (IV) Catheterization Hand
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
IV-M-001-M IV-M-001-N IV-M-001-B
470336-958EA 276.45 USD InStock
470336-958 470336-960 470336-956
Medicor® Peripheral Intravenous (IV) Catheterization Hand
Models Human Biology Models
Easily practice small vein phlebotomy.
  • Available In three skin tones
  • Mold resistant
  • Self healing skin to 22G
Made entirely of proprietary MedicSkin, this lifelike hand provides students with a realistic experience for practicing peripheral IV insertion and phlebotomy procedures in the small veins of the hand. Students can practice on the dorsal, basilic, or cephalic veins. Track marks heal quickly, allowing students to practice repeatedly.
Ordering information: Skin and veins are not replaceable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty