Would you like to log in again?
Medicor® Peripheral Intravenous (IV) Catheterization Arm Model
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
IV-BT-001-TC-M IV-BT-001-TC-N IV-BT-001-TC-B
470337-041EA 860.7 USD InStock
470337-041 470337-042 470336-962
Medicor® Peripheral Intravenous (IV) Catheterization Arm Model
Models Human Biology Models
Perform IV and IM on a single model.
  • Available in three skin tones
  • Mold resistant
  • Self healing skin to 22G
A best-selling product, this IV arm features four veins to practice performing peripheral IV insertions. Each arm comes with two 1000 ml bags to use for simulating an IV drip and blood flow through the arm. After inserting the catheter correctly, students experience a realistic flashback for placement confirmation.
Caution: Some components of this product may contain latex.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty