Would you like to log in again?
Medicor® Sticky Wounds
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
P-PCBPK4-001-N P-OG-001-M P-PC-001-6-M P-OG-001-B P-PC-001-6-N P-OG-001-N P-PSCK4-001-N P-PC-001-6-B P-SC-001-B P-SC-001-M P-SPC-001-M P-PSCK4-001-B P-PCBPK4-001-B P-SC-001-N P-PSCK4-001-M P-SPC-001-B P-SPC-001-N P-PCBPK4-001-M
470336-982PK 554.85 USD InStock
470336-982 470336-980 470337-014 470337-025 470337-015 470337-026 470337-016 470337-027 470337-028 470337-023 470337-024 470336-996 470336-986 470336-994 470336-984 470336-978 470336-998 470336-976
Medicor® Sticky Wounds
Models Human Biology Models
Add to manikins or standard patients.
  • Easy to remove and reuse
  • Expand training with existing manikins
  • Skin tone colors match Medicor and CAE
Medicor sticky wounds are made entirely of MedicGel for a lifelike appearance. Able to stick to any surface without adhesive, these wounds can be applied to any manikin or standardized patient. All sticky wounds are stain-resistant and hypoallergenic.
TIP: If the product is not sticking well due to dust or other contaminants, rinse with tap water to reactivate the material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty