Would you like to log in again?
Medicor® Stop The Bleed Arm
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
MBMSTB001SMM MBMSTB001SMB MBMSTB001SMN
470336-992EA 2560.9 USD InStock
470336-992 470336-990 470336-988
Medicor® Stop The Bleed Arm
Models Human Biology Models
Learn trauma response methods.
  • Available in three skin tones
  • Two wounds for practice
Challenge students' simulation scenarios with this innovative product made entirely of proprietary MedicSkin. Experience realistic lower-arm bleeding that stops when a tourniquet is properly applied. Continue the challenge by initiating bleeding on the upper-arm wound, forcing students to use critical thinking skills to find the second source of bleeding.
Note: This kit was not developed as part of the Stop the Bleed® Program, and is not endorsed by it.
Ordering information: Some items in images may not be included.
Caution: Some components of this product may contain latex.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty