Would you like to log in again?
Medicor® Infant Foot For Heel Prick Screening Test
Supplier: ELEVATE HEALTHCARE, INC. SE
Medicor®
M-NNPES-001-B M-NNPES-001-N M-NNPES-001-M
470337-035EA 250.4 USD InStock
470337-035 470337-036 470337-037
Medicor® Infant Foot For Heel Prick Screening Test
Models Human Biology Models
The only heel prick trainer available.
  • Contains internal reservoir
  • Available in three skin tones
Performing infant blood screenings is crucial to detect rare but serious disorders that may affect a baby's health. Students can learn and practice the appropriate technique using this infant foot that features lifelike blood sampling. The model realistically simulates the softness and structure of a baby's heel while the internal reservoir releases a small trickle of blood with the application of light pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty