Would you like to log in again?
CRISPR Adaptive Immunity
Supplier: 3-D Molecular Designs
CSCIAK-01
470337-146CS 780.35 USD InStock
470337-146
CRISPR Adaptive Immunity
Educational Materials Biology Educational Materials AP Biology Learning Activities
What does CRISPR do?
  • Interactive modeling
  • How CRISPR was discovered and what it does
  • Reusable
Discover how bacteria remember past viral infections and activate their CRISPR systems to search for and destroy the remembered virus’ DNA - preventing infection. This introduction to CRISPR biology will prepare your students to use making the cut with CRISPR-Cas9 and understand how the Cas9 protein works.
With the CRISPR Adaptive Immunity Kit's engaging foam pieces, your students can:
Build the bacteria’s CRISPR array which includes Cas9, trace, spacer and repeats.
Detect the presence of viral double-stranded DNA.
Integrate a short fragment of this viral genome into the bacteria’s CRISPR array.
Express the CRISPR array to create the Cas9 proteins and the guide RNA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty